missing translation for 'onlineSavingsMsg'
Learn More

Tuning Solution 4 For ICP, SPEX CertiPrep™

Catalog No. CLTUNE4
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLTUNE4 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLTUNE4 Supplier SPEX CertiPrep Group LLC Supplier No. CLTUNE4
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Co10μg/mL
In10μg/mL
Li10μg/mL
Tl10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Tuning Solution 4
Packaging Triple-leached LDPE bottle
Name Note For ICP/MS
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Co,In,Li
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Element Co, In, Li
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade PPT
Concentration or Composition (by Analyte or Components) 10mg/L
Show More Show Less

Product intended for laboratory use only