missing translation for 'onlineSavingsMsg'
Learn More

Spike Sample Standard 3 For ICP, SPEX CertiPrep™

Catalog No. CLSPIKE3
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLSPIKE3 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLSPIKE3 Supplier SPEX CertiPrep Group LLC Supplier No. CLSPIKE3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al200μg/mL
Ba200μg/mL
Co50μg/mL
Mn50μg/mL
Ni50μg/mL
V50μg/mL
Zn50μg/mL
Cu25μg/mL
Cr20μg/mL
Sb10μg/mL
Ag5μg/mL
Be5μg/mL
Cd5μg/mL
Tl5μg/mL
As4μg/mL
Pb2μg/mL
Se1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Spike Sample 3
Packaging Triple-leached LDPE bottle
Name Note For ILM05.2 with ICP-MS
CAS 7440-02-0, 7732-18-5, 7440-39-3, 7429-90-5, 7439-96-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 93.13, 0.9, 0.02, 0.02, 0.0, 0.0
Analyte or Component Names Al (200mg/L),Ba (200mg/L),Co (50mg/L)
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Element Al, Ba, Co
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Grade PPT
Show More Show Less

Product intended for laboratory use only