missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element Solution For ICP, SPEX CertiPrep™

Numéro de catalogue. CLMSSETN
Change view
Click to view available options
Quantity:
7 x 125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
CLMSSETN 7 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CLMSSETN Fournisseur SPEX CertiPrep Group LLC Code fournisseur CLMSSETN
Il en reste null

Comprend

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Multi-Element Solutions Standards Set for ICP-MS, 125 mL, Set of 7, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • 125 mL Volume
  • Standards Set for ICP-MS

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Spécifications

Chemical Name or Material Multi-Element Solutions without Mercury
Packaging Triple-leached LDPE bottle
Name Note For ICP/MS; Without mercury
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Claritas PPT Grade ASTM I Water Blank,Claritas PPT Grade Hydrochloric Acid Blank,Claritas PPT Grade Nitric Acid Blank
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Element Water, Hydrochloric Acid, Nitric Acid
Solvent or Matrix H2O; 2% HCl; 2% HNO3; 5% HNO3; 10% HCl/1% HNO3; H2O/Tr. HF/Tr. HNO3
Quantity 7 x 125 mL
Afficher plus de résultats Afficher moins de résultats

Product intended for laboratory use only