missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CKAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APSA6835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APSA6835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APSA6835 Supplier Invitrogen™ Supplier No. PA316835
Only null left

Rabbit Polyclonal Antibody

CKAP5 Polyclonal Antibody for Western Blot, ICC/IF, IP

Colonic and hepatic tumor over-expressed protein (ch-TOG) belongs to the TOG/XMAP215/Dis1 evolutionarily ancient family of microtubule-associated proteins with isoforms found in all major kingdoms of eukaryotes. This protein is required for the assembly and function of the meiotic or mitotic spindles during cell division and is localized to microtubule-organizing centers (MTOCs) during interphase or to the spindle poles and centrosomes during mitosis or cytokinesis.

Specifications

Antigen CKAP5
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene CKAP5
Gene Accession No. A2AGT5, Q14008
Gene Alias 3110043H24Rik; 4930432B04Rik; CHTOG; ch-TOG; ch-TOGp; CKAP5; colonic and hepatic tumor overexpressed gene protein; colonic and hepatic tumor over-expressed gene protein; colonic and hepatic tumor over-expressed protein; cytoskeleton associated protein 5; cytoskeleton-associated protein 5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp
Gene Symbols CKAP5
Host Species Rabbit
Immunogen A recombinant segment of the C-terminal domain of human ch-TOG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311191, 75786, 9793
Target Species Human, Mouse, Rat, Mammalia
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less