missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CK2 alpha-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595701 Supplier Invitrogen™ Supplier No. PA595701
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: LGTEELYGYL KKYHIDLDPH FNDILGQHSR KRWENFIHSE NRHLVSPEAL DLLDKLLRYD HQQRLTAKEA MEHPYFYPVV KEQSQPCADN AVLSSGLTAA R Positive Samples: K-562, 293T, HeLa, Mouse testis, Mouse brain.

Casein kinase II is a protein serine/threonine kinase that is thought to be present in all eukarotic cells, implying that it has fundamental cellular functions. The holoenzyme is a tetramer containing 2 alpha or alpha-prime subunits (or one of each) and 2 beta subunits. The function of beta subunits is unknown and the alpha subunit is the catalytic unit. CK2 functions as a tetrameric complex consisting of two regulatory beta-subunits and two catalytic units (alpha and alpha') in a homomeric or heteromeric conformation. Whilst the alpha- and alpha'-subunits are catalytically identical, proteins that regulate CK2, such as cdc2 and Hsp90, preferentially bind to the alpha and not the alpha'-subunit. CK2 can phosphorylate a number of key intracellular signaling proteins implicated in tumor suppression (p53 and PTEN) and tumorigenesis (myc, jun, NF-kappaB). CK2 is also thought to influence Wnt signaling via beta-catenin phosphorylation and the PI 3-K signaling pathway via the phosphorylation of Akt.

Specifications

Antigen CK2 alpha-2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CSNK2A2
Gene Accession No. O54833, P19784
Gene Alias 1110035J23Rik; C77789; casein kinase 2 alpha; casein kinase 2 alpha'; casein kinase 2 alpha 2; casein kinase 2, alpha prime polypeptide; casein kinase II subunit alpha'; casein kinase II, alpha 2, polypeptide; CK II alpha'; CK2; CK2A2; CK2alpha'; CSNK2A1; CSNK2A2
Gene Symbols CSNK2A2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13000, 1459, 307641
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less