missing translation for 'onlineSavingsMsg'
Learn More

Medisca Citrulline (L) (Dietary Supplement Grade)

Catalog No. 14185963
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185963 500 g
14186159 100 g
14185470 25 g
14186238 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14185963 Supplier Medisca Supplier No. 118608

Citrulline (L) (Dietary Supplement Grade) is white and comes as crystals or as a crystalline powder. It is practically odorless.

Citrulline (L) is white and comes as crystals or as a crystalline powder. It is practically odorless.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Citrulline

Specifications

Chemical Name or Material Citrulline (L) (Dietary Supplement Grade)
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 372-75-8
Type Drug Standard
Molecular Weight (g/mol) 175.187
Quantity 500 g
Percent Purity 99.5 to 101.0 %
Grade Ungraded