missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citronellyl acetate, ≥95.0% (GC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 150-84-5 |
| Density | 0.891 g/mL (at 25°C) |
| Flash Point | 103°C |
| Physical Form | Neat |
| Linear Formula | CH3 CO2 CH2 CH2 CH(CH3)CH2 CH2 CH=C(CH3)2 |
| MDL Number | MFCD00015039 |
| Boiling Point | 240°C |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 198.30 |
| Show More |