missing translation for 'onlineSavingsMsg'
Learn More

(-)-β-Citronellol, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111015437
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015437 5 mL
111015436 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015437 Supplier Merck Emd Millipore Supplier No. 274835MLF

Citronellol is a monoterpene alcohol found in essential oils of Cymbopogon winterianus Jowitt. S-(−)-isomer that is less common. It is prevalent in geranium and citronella oils.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 7540-51-4
Density 0.854 g/mL (at 20°C)
Flash Point 99°C - Closed Cup
Physical Form Neat
Linear Formula C10H20 O
MDL Number MFCD00063214
Boiling Point 221°C to 224°C
Synonym beta-Rhodinol; (S)-(-)-beta-Citronellol; (S)-3,7-Dimethyl-6-octen-1-ol
Type Analytical Standard
Quantity 5 mL
Formula Weight 156.27
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less