missing translation for 'onlineSavingsMsg'
Learn More

Citronellic Acid 95.0+%, TCI America™

Catalog No. C170825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C170825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C170825ML Supplier TCI America Supplier No. C170825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 502-47-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00002728
InChI Key GJWSUKYXUMVMGX-UHFFFAOYNA-N
Synonym citronellic acid, 3,7-dimethyl-6-octenoic acid, rhodinolic acid, citronellate, rhodinic acid, 6-octenoic acid, 3,7-dimethyl, fema no. 3142, citronellicacid, callitrol, acmc-20apjr
PubChem CID 10402
ChEBI CHEBI:80507
IUPAC Name 3,7-dimethyloct-6-enoic acid
SMILES CC(CCC=C(C)C)CC(O)=O

Specifications

Color Yellow
Quantity 25 mL
UN Number 2810
Formula Weight 170.25
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Citronellic Acid
EINECSNumber (2)-0609
RTECSNumber RH3230000
TSCA Yes