accessibility menu, dialog, popup

UserName

CAS RN 502-47-6

IUPAC Name: 3,7-dimethyloct-6-enoic acid
Synonyms: citronellic acid citronellate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: GJWSUKYXUMVMGX-UHFFFAOYNA-N
SMILES: CC(CCC=C(C)C)CC(O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)