missing translation for 'onlineSavingsMsg'
Learn More

Citronellic acid, 94%

Catalog No. p-7026703
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2286214 25 g
AAB2286222 100 g
AAB2286236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2286214 Supplier Thermo Scientific Chemicals Supplier No. B2286214
Only null left

CAS: 502-47-6 | C10H18O2 | 170.25 g/mol

Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 502-47-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00002728
InChI Key GJWSUKYXUMVMGX-UHFFFAOYNA-N
Synonym citronellic acid, 3,7-dimethyl-6-octenoic acid, rhodinolic acid, citronellate, rhodinic acid, 6-octenoic acid, 3,7-dimethyl, fema no. 3142, citronellicacid, callitrol, acmc-20apjr
PubChem CID 10402
ChEBI CHEBI:80507
IUPAC Name 3,7-dimethyloct-6-enoic acid
SMILES CC(CCC=C(C)C)CC(O)=O

Specifications

Density 0.923
Boiling Point 121°C to 122°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4535
Quantity 25 g
UN Number UN2810
Solubility Information Insoluble in water.
Formula Weight 170.25
Percent Purity 94%
Chemical Name or Material Citronellic acid
Hazard Category H312
Hazard Statement GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P302+P352-P312-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-939-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only