missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Citronellal 96.0+%, TCI America™
$121.16 - $344.86
Chemical Identifiers
| CAS | 5949-05-3 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.25 |
| MDL Number | MFCD00075588 |
| InChI Key | NEHNMFOYXAPHSD-JTQLQIEISA-N |
| Synonym | s---citronellal, --citronellal, s-citronellal, s-3,7-dimethyloct-6-enal, 3s-3,7-dimethyl-6-octenal, 3s---citronellal, s---3,7-dimethyl-6-octenal, s-3,7-dimethyl-6-octenal, 3s-3,7-dimethyloct-6-enal, 6-octenal, 3,7-dimethyl-, 3s |
| PubChem CID | 443157 |
| ChEBI | CHEBI:368 |
| IUPAC Name | (3S)-3,7-dimethyloct-6-enal |
| SMILES | CC(CCC=C(C)C)CC=O |
Chemical Identifiers
| 5949-05-3 | |
| 154.25 | |
| NEHNMFOYXAPHSD-JTQLQIEISA-N | |
| 443157 | |
| (3S)-3,7-dimethyloct-6-enal |
| C10H18O | |
| MFCD00075588 | |
| s---citronellal, --citronellal, s-citronellal, s-3,7-dimethyloct-6-enal, 3s-3,7-dimethyl-6-octenal, 3s---citronellal, s---3,7-dimethyl-6-octenal, s-3,7-dimethyl-6-octenal, 3s-3,7-dimethyloct-6-enal, 6-octenal, 3,7-dimethyl-, 3s | |
| CHEBI:368 | |
| CC(CCC=C(C)C)CC=O |
Specifications
| 5949-05-3 | |
| C10H18O | |
| 5 mL | |
| NEHNMFOYXAPHSD-JTQLQIEISA-N | |
| (3S)-3,7-dimethyloct-6-enal | |
| 443157 | |
| 154.25 | |
| Liquid |
| Colorless | |
| MFCD00075588 | |
| s---citronellal, --citronellal, s-citronellal, s-3,7-dimethyloct-6-enal, 3s-3,7-dimethyl-6-octenal, 3s---citronellal, s---3,7-dimethyl-6-octenal, s-3,7-dimethyl-6-octenal, 3s-3,7-dimethyloct-6-enal, 6-octenal, 3,7-dimethyl-, 3s | |
| CC(CCC=C(C)C)CC=O | |
| 154.25 | |
| CHEBI:368 | |
| ≥96.0% (GC) | |
| (-)-Citronellal |
Safety and Handling
EINECSNumber : (2)-0514
TSCA : Yes