accessibility menu, dialog, popup

UserName

CH3(S)H3CCH3O

CAS RN 5949-05-3

IUPAC Name: (3S)-3,7-dimethyloct-6-enal
Synonyms: (S)-(-)-citronellal (3S)-(-)-citronellal
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: NEHNMFOYXAPHSD-JTQLQIEISA-N
SMILES: C[C@@H](CCC=C(C)C)CC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results