missing translation for 'onlineSavingsMsg'
Learn More

(-)-Citronellal 96.0+%, TCI America™

Catalog No. C14545ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14545ML 5 mL
C145425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14545ML Supplier TCI America Supplier No. C14545ML
Only null left

Chemical Identifiers

CAS 5949-05-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00075588
InChI Key NEHNMFOYXAPHSD-JTQLQIEISA-N
Synonym s---citronellal, --citronellal, s-citronellal, s-3,7-dimethyloct-6-enal, 3s-3,7-dimethyl-6-octenal, 3s---citronellal, s---3,7-dimethyl-6-octenal, s-3,7-dimethyl-6-octenal, 3s-3,7-dimethyloct-6-enal, 6-octenal, 3,7-dimethyl-, 3s
PubChem CID 443157
ChEBI CHEBI:368
IUPAC Name (3S)-3,7-dimethyloct-6-enal
SMILES CC(CCC=C(C)C)CC=O

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Citronellal
EINECSNumber (2)-0514
TSCA Yes