missing translation for 'onlineSavingsMsg'
Learn More

Citronellal, 95+%

Catalog No. p-93327
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405291000 100 g Glass bottle
AC405295000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405291000 Supplier Thermo Scientific Chemicals Supplier No. 405291000
Only null left

CAS: 106-23-0 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-23-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00038090
InChI Key NEHNMFOYXAPHSD-SNVBAGLBSA-N
Synonym citronellal, rhodinal, 3,7-dimethyl-6-octenal, 2,3-dihydrocitral, 6-octenal, 3,7-dimethyl, citronellel, citronella, 3,7-dimethyl-6-octen-1-al, beta-citronellal, d-rhodinal
PubChem CID 7794
ChEBI CHEBI:47856
IUPAC Name 3,7-dimethyloct-6-enal
SMILES C[C@H](CCC=C(C)C)CC=O

Specifications

Color Yellow
Density 0.8600g/mL
Boiling Point 206°C to 208°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.45
Linear Formula (CH3)2CCHCH2CH2CH(CH3)CH2CHO
Quantity 100 g
Specific Gravity 0.86
Merck Index 15, 2328
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents,ethanol,ether
Formula Weight 154.25
Percent Purity 93%
Physical Form Liquid
Chemical Name or Material Citronellal, pract., 93%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation or rash occurs: G
EINECSNumber 203-376-6
RTECSNumber RH2140000
TSCA TSCA

RUO – Research Use Only