missing translation for 'onlineSavingsMsg'
Learn More

(±)-Citronellal, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019479
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019479 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019479 Supplier Merck Emd Millipore Supplier No. 726381ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical standard used for research applications in flavors, fragrances, and personal care formulations.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Chemical Name or Material Citronellal
CAS 106-23-0
Density 0.857 g/mL (at 25°C)
Flash Point 86°C - Closed Cup
Physical Form Neat
Linear Formula (CH3)2 C=CHCH2 CH2 CH(CH3)CH2 CHO
Technique Gas Chromatography
MDL Number MFCD00038090
Boiling Point 207°C
Synonym (+/-)-3,7-Dimethyl-6-octenal
Type Analytical Standard
Industry Type Flavors and Fragrances
Molecular Weight (g/mol) 154.25
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Show More Show Less