missing translation for 'onlineSavingsMsg'
Learn More

Citrinin Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111017990
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017990 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017990 Supplier Merck Emd Millipore Supplier No. 354101ML
Only null left

Citrinin is a hepato-nephrotoxic mycotoxin. It can be produced by several filamentous fungi genera of Monauscus, Aspergillus, and Penicillium, which is commonly found as a contaminant in stored grains, foods, feedstuffs and also in biological fluids.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 518-75-2
Flash Point 2.0°C - Closed Cup
Linear Formula C13H14O5
MDL Number MFCD00006912
Synonym 4,6-Dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3 H-2-benzopyran-7-carboxylic acid solution
Type Analytical Standard
Quantity 1 mL
Formula Weight 250.25
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile