missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric/Phosphate Buffer, McIlvaine, pH 3.20, Ricca Chemical
$175.13 - $175.13
Specifications
| Chemical Name or Material | Citric/Phosphate Buffer |
|---|---|
| Name Note | McIlvaine, pH 3.20 |
| CAS Min % | 98.0,0.09,0.03 |
| CAS Max % | 100.0,0.09,0.03 |
| Color | Colorless |
Specifications
| Citric/Phosphate Buffer | |
| 98.0,0.09,0.03 | |
| Colorless | |
| 1X | |
| 3.2 | |
| Buffer |
| McIlvaine, pH 3.20 | |
| 100.0,0.09,0.03 | |
| 77-92-9,7558-79-4 | |
| Water Analysis | |
| Liquid | |
| Laboratory |