missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Purified, Reagents

Catalog No. 01337606
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337606 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337606 Supplier Labchem Inc Supplier No. C21045003A5
Only null left

Citric Acid Monohydrate, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Color Colorless/White
Grade Reagent
Quantity 3 kg
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.542
Chemical Name or Material Citric Acid Monohydrate, Purified