missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Purified, Reagents

Citric Acid Monohydrate, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc C2104500500A5

 View more versions of this product

Catalog No. 01-337-605

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Citric Acid Monohydrate, Purified
5949-29-1
C6H10O8
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Reagent
Poly Bottle
1.542
153°C
Colorless/White
2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
210.14
500 g
Solid

Chemical Identifiers

5949-29-1
210.14
2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
C6H10O8
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions