missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Honeywell Fluka™

Catalog No. p-7071954
Change view
Click to view available options
Quantity:
1 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002248 1 kg
6002249 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002248 Supplier Honeywell Chemicals Supplier No. 274911KG
Only null left

Tested according to European Pharmacopoeia

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

Linear Formula HOC(COOH)(CH2COOH)2 · H2O
UN Number NONH for all modes of transport
Beilstein 4018641
Solubility Information Soluble in ethyl acetate (5.28g/100g), Amyl Acetate(5.98g/100g), diethyl ether(2.17g/100g), methanol(197g/100g), chloroform(0.007g/100 g)
Formula Weight 210.14g/mol
Quantity 1 kg
Flash Point 173°C (343.4°F)
Chemical Name or Material Citric Acid Monohydrate, Tested according to Ph.Eur.
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 201-069-1
RTECSNumber GE7810000