missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate (Granular/USP), Fisher Chemical™

Catalog No. p-92734
Change view
Click to view available options
Quantity:
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A1103 3 kg Poly Bottle
A11010 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1103 Supplier Thermo Fisher Scientific Supplier No. FLA1103
Only null left

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

pH 2.2
Melting Point 135°C to 152°C
Boiling Point 175°C
Color White
Formula Weight 210.14
Percent Purity 99.5 to 100.5% (USP)
Grade USP
Density 1.54g/mL
Quantity 3 kg
Flash Point 174°C
Ignition Residue 0.1% max. (USP)
Packaging Poly Bottle
Physical Form Solid
Chemical Name or Material Citric Acid Monohydrate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Risk of serious damage to eyes. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0