missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Monohydrate, Granular, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7157271
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18601729 500 g Poly Bottle
18601730 2.5 kg Poly Bottle
18601731 12 kg Poly Pail
18601732 50 kg Fiber Drum
4 options

Catalog No. 18601729

Supplier: Spectrum Chemical Mfg Cor CI132500GM

Only null left

CI132, 5949-29-1, C6H8O7·H2O

Spectrum™ Chemical Citric Acid, Monohydrate, Granular, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Percent Purity 99.5 to 100.5%
Grade USP
Quantity 500 g
Ignition Residue 0.1%
Packaging Poly Bottle