missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate (Granular/Certified ACS), Fisher Chemical™

Catalog No. A1043
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A1043 3 kg Poly Bottle
A104500 500 g Poly Bottle
A104250LB 250 lb. Fiber Drum
A10410 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1043 Supplier Thermo Fisher Scientific Supplier No. FLA1043
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

pH 2.2
Melting Point 135°C to 152°C
Boiling Point 175°C
Color White
Assay Percent Range 99.0 to 102.0 %
Linear Formula (HO2CCH2)2C(OH)(CO2H)·H2O
Formula Weight 210.14
Percent Purity 99.0 to 102.0%
Grade Certified ACS
Density 1.54g/mL
Quantity 3 kg
Flash Point 174°C
Ignition Residue 0.02% max.
Packaging Poly Bottle
Physical Form Solid
Chemical Name or Material Citric Acid Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Risk of serious damage to eyes. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0