missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, For Analysis, ACS, EMSURE™, MilliporeSigma™

Catalog No. p-7082905
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1002440500 500 g Plastic Bottle
M1002441000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1002440500 Supplier Technidata Supplier No. 1.00244.0500
Only null left

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

pH 1.85
Melting Point 135°C to 152°C
Color White
Solubility Information 880g/L
Formula Weight 210.14
Percent Purity 99.5 to 100.5 %
Grade ACS/ISO/Reagent Ph. Eur.
Density 1.54 g/3 (20 °C)
Quantity 500 g
Packaging Plastic Bottle
Physical Form Solid
Vapor Pressure 0.1 hPa at 20°C
Show More Show Less
EINECSNumber 201-069-1