missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Monohydrate, Crystalline Powder, USP, 99.5-100.5%, Spectrum™ Chemical

C1296, 5949-29-1, C6H8O7·H2O

Supplier:  Spectrum Chemical Mfg Cor C129650KGBL

 View more versions of this product

Catalog No. 18601724

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Citric Acid, Monohydrate, Crystalline Powder, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

5949-29-1
C6H10O8
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
210.14
USP
0.001
1
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
99.5 to 100.5%
50 kg
Fiber Drum

Chemical Identifiers

5949-29-1
210.14
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
C6H10O8
YASYEJJMZJALEJ-UHFFFAOYSA-N
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions