missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Monohydrate, Crystalline Powder, USP, 99.5-100.5%, Spectrum™ Chemical
C1296, 5949-29-1, C6H8O7·H2O
Supplier: Spectrum Chemical Mfg Cor C129625KG
Description
Spectrum™ Chemical Citric Acid, Monohydrate, Crystalline Powder, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 5949-29-1 | |
| C6H10O8 | |
| O.OC(=O)CC(O)(CC(O)=O)C(O)=O | |
| 210.14 | |
| USP | |
| 0.001 |
| 1 | |
| YASYEJJMZJALEJ-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate | |
| 99.5 to 100.5% | |
| 2.5 kg | |
| Poly Bottle |
Chemical Identifiers
| 5949-29-1 | |
| 210.14 | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
| C6H10O8 | |
| YASYEJJMZJALEJ-UHFFFAOYSA-N | |
| O.OC(=O)CC(O)(CC(O)=O)C(O)=O |