missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Monohydrate, Crystalline Powder, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18601721
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18601721 500 g Poly Bottle
18601722 2.5 kg Poly Bottle
18601723 12 kg Poly Pail
18601724 50 kg Fiber Drum
4 options

Catalog No. 18601721

Supplier: Spectrum Chemical Mfg Cor C1296500GM

Only null left

C1296, 5949-29-1, C6H8O7·H2O

Spectrum™ Chemical Citric Acid, Monohydrate, Crystalline Powder, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Max % 1
Percent Purity 99.5 to 100.5%
Grade USP
Quantity 500 g
Ignition Residue 0.001
Packaging Poly Bottle