missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Monohydrate, Crystal, ACS, 99.0-102.0%, Spectrum™ Chemical

Catalog No. p-7165331
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609815 25 g Poly Bottle
18609816 500 g Poly Bottle
18609817 2.5 kg Poly Bottle
18609818 12 kg Poly Pail
18609819 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609815 Supplier Spectrum Chemical Mfg Cor Supplier No. C128525GM
Only null left

Chemical Identifiers

CAS 5949-29-1

Specifications

CAS Max % 1
Percent Purity 99 to 102%
Grade ACS
Quantity 25 g
Packaging Poly Bottle
Residue after Ignition 0.0002

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.