missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, For Radium, Certified, 1.00M ±0.01M (3.0N), LabChem™

Catalog No. p-5388372
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
500 mL
Catalog No. Quantity
LC131801 500 mL
LC131802 1 L
2 options

Catalog No. LC131801

Supplier: LabChem LC131801

Only null left

Chemical Identifiers

CAS 77-92-9 , 7732-18-5
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Color Colorless
CAS Max % 0.808, 0.192
Linear Formula C6H8O7
Solubility Information Soluble in water
Concentration 1.00M ±0.01M (3.0N)
Formula Weight 192.12
Grade Certified
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Traceability to NIST Traceable to NIST
Chemical Name or Material Citric Acid, For Radium
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature