missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, Electrophoresis Grade, 99.5+%

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol

Supplier:  Thermo Scientific Chemicals J64322A3

 View more versions of this product

Catalog No. AAJ64322A3


Only null left
Explore available promotions

Description

Description

Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.

Solubility
Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications

Specifications

Citric acid
153°C to 154°C
White
MFCD00011669
14,2326
Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene.
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
CHEBI:30769
≥99.5%
1.665
Odorless
Hygroscopic
1.6
77-92-9
C6H8O7
782061
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.13
Electrophoresis
2 kg
Powder

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335-H500

EINECSNumber : 201-069-1

RTECSNumber : GE7350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only