missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, Electrophoresis Grade, 99.5+%

Catalog No. p-7042912
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6432222 100 g
AAJ6432236 500 g
AAJ64322A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6432222 Supplier Thermo Scientific Chemicals Supplier No. J6432222

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol

Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is a starting point in the Krebs cycle and is a key intermediate in metabolism. It has anticoagulant activity; as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot. It may be used to adjust pH and as a sequestering agent for the removal of trace metals.

Solubility
Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1.6
Melting Point 153°C to 154°C
Color White
Beilstein 782061
Merck Index 14,2326
Solubility Information Soluble in water. Also soluble in alcohol and methanol. Insoluble in chloroform and benzene.
Formula Weight 192.13
Percent Purity ≥99.5%
Grade Electrophoresis
Density 1.665
Quantity 100 g
Odor Odorless
Physical Form Powder
Sensitivity Hygroscopic
Chemical Name or Material Citric acid
Show More Show Less
Hazard Category H319-H335-H500
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 201-069-1
RTECSNumber GE7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only