missing translation for 'onlineSavingsMsg'
Learn More

Citric acid diammonium salt, 99+%

Catalog No. aa3322136
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3322136 500 g
AA3322122 100 g
AA33221A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3322136 Supplier Thermo Scientific Chemicals Supplier No. 03322136
Only null left

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol

Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3012-65-5
Molecular Formula C6H14N2O7
Molecular Weight (g/mol) 226.19
MDL Number MFCD00013068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
Synonym ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
PubChem CID 13710713
ChEBI CHEBI:63076
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Specifications

Color White
Density 1.48 g/mL
Odor Odorless
Linear Formula HOC(CO2H)(CH2CO2NH4)2
Quantity 500 g
Merck Index 14512
Solubility Information Soluble in water. Slightiy soluble in alcohol.
Formula Weight 226.19
Percent Purity ≥99%
Physical Form Granules
Chemical Name or Material Citric acid diammonium salt
Show More Show Less
Hazard Category H319-H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 221-146-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only