accessibility menu, dialog, popup

UserName

CAS RN 3012-65-5

IUPAC Name: diammonium 3-carboxy-3-hydroxypentanedioate
Synonyms: microstop ammonium citrate, dibasic citric acid diammonium salt ammonium hydrogen citrate ammonium monohydrogen citrate diammonium citrate ammonium monohydrogencitrate dibasic ammonium citrate ammonium citrate dibasic diammonium hydrogen citrate
Molecular Weight (g/mol): 226.19
Molecular Formula: C6H14N2O7
InChi Key: YXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
Grade 
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (5)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (5)
  • (14)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (6)