missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, diammonium salt, 99+%, for biochemistry, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 448025000

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC448025000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3012-65-5
226.19
YXVFQADLFFNVDS-UHFFFAOYSA-N
13710713
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
C6H14N2O7
MFCD00013068
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
CHEBI:63076
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Specifications

Citric acid, diammonium salt
1.5000g/mL
Plastic bottle
MFCD00013068
15,509
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
YXVFQADLFFNVDS-UHFFFAOYSA-N
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
13710713
226.19
Biochemistry
3012-65-5
99+%
C6H14N2O7
500g
1.5
Solubility in water: soluble
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
226.19
CHEBI:63076
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 221-146-3

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...