missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Anhydrous, Purified, Reagents
Citric Acid Anhydrous, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C21045501KG
Specifications
| Citric Acid Anhydrous, Purified | |
| 77-92-9 | |
| Colorless/White | |
| 2-hydroxypropane-1,2,3-tricarboxylic Acid | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O | |
| 192.12 | |
| 1 kg | |
| Solid |
| 153°C | |
| 175°C | |
| C6H8O7 | |
| KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid | |
| Pure | |
| Poly Bottle | |
| 1.665 |
Chemical Identifiers
| 77-92-9 | |
| 192.12 | |
| 2-hydroxypropane-1,2,3-tricarboxylic Acid | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O |
| C6H8O7 | |
| KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid |