missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Anhydrous, Purified, Reagents

Catalog No. p-200021214
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337604 10 kg
01337603 1 kg
01337602 3 kg
01337601 500 g
01337600 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337604 Supplier Labchem Inc Supplier No. C210455010KG
Only null left

Citric Acid Anhydrous, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Boiling Point 175°C
Color Colorless/White
Grade Pure
Quantity 10 kg
Packaging Poly Pail
Physical Form Solid
Specific Gravity 1.665
Chemical Name or Material Citric Acid Anhydrous, Purified