missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Anhydrous (Crystalline/USP), Fisher Chemical™

$710.14 - $710.14

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A95-3
SDS View Documents Promotion Details
Fisher Chemical
FLA953
3 kg Poly Bottle
Each for $710.14
Case of 4 Each for $2,840.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

153°C
175°C
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.13
USP
3 kg
Poly Bottle
Citric Acid, Anhydrous
77-92-9
C6H8O7
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
CHEBI:30769
99.5 to 100.5% (USP)
1.67g/mL
0.1% max. (USP)
Crystalline
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'