missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Anhydrous (Crystalline/USP), Fisher Chemical™

Supplier:  Fisher Chemical FLA953

 View more versions of this product

Catalog No. A95-3



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Citric Acid, Anhydrous
77-92-9
C6H8O7
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
CHEBI:30769
99.5 to 100.5% (USP)
1.67g/mL
0.1% max. (USP)
Crystalline
153°C
175°C
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.13
USP
3 kg
Poly Bottle

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions