missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, ≥99.5%, ACS reagent, anhydrous
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Supplier: Thermo Scientific Chemicals 423565000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Citric acid | |
| 77-92-9 | |
| HO2CCH2C(OH)(CO2H)CH2CO2H | |
| citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
| KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 311 | |
| 192.13 | |
| ACS Reagent | |
| 100°C |
| 153.0°C | |
| C6H8O7 | |
| MFCD00011669 | |
| Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O | |
| 192.12 | |
| CHEBI:30769 | |
| 99.6% | |
| 500 g |
Chemical Identifiers
| 77-92-9 | |
| 192.12 | |
| KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
| 311 | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid |
| C6H8O7 | |
| MFCD00011669 | |
| citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
| CHEBI:30769 | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O |
RUO – Research Use Only