missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, 20% w/v, Reagents
Specifications
Specifications
| pH | <2 |
| Boiling Point | 100°C |
| Color | Colorless |
| Quantity | 20 L |
| Packaging | Cubitainer |
| Physical Form | Liquid |
| Specific Gravity | 1.0808 |
| Percent Purity | 20% |
| Grade | Reagent |
| Chemical Name or Material | Citric Acid, 20% w/v |