missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 10% w/v, Reagents

Catalog No. 01335603
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335603 1 L
01335602 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335603 Supplier Labchem Inc Supplier No. CC2041001A
Only null left

Citric Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 77-92-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
IUPAC Name water
SMILES O

Specifications

pH 1.72
Boiling Point 101°C
Color Colorless
Percent Purity 10% w/v
Grade Reagent
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0405
Chemical Name or Material Citric Acid, 10% w/v