missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 10% (w/v) from USP grade material, Ricca Chemical

Catalog No. p-3251505
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
212016 500 mL Natural Poly Bottle
212032 1 L Natural Poly Bottle
RC21201 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 212016 Supplier Ricca Chemical Company Supplier No. 212016
Only null left

Chemical Identifiers

CAS 7732-18-5 , 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Min % 89.0, 9.0
Color Colorless
CAS Max % 93.0, 11.0
Concentration 10% (w/v)
Percent Purity 10% w/v
Grade Laboratory
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Citric Acid