missing translation for 'onlineSavingsMsg'
Learn More

Citrazinic Acid 96.0+%, TCI America™

Catalog No. p-7124503
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C057125G 25 g
C0571500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C057125G Supplier TCI America Supplier No. C057125G

Chemical Identifiers

CAS 99-11-6
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00006274
InChI Key CSGQJHQYWJLPKY-UHFFFAOYSA-N
Synonym citrazinic acid, 2,6-dihydroxyisonicotinic acid, 2,6-dihydroxypyridine-4-carboxylic acid, 2,6-dihydroxy-4-carboxypyridine, 2,6-dihydroxy-4-pyridinecarboxylic acid, kyselina citrazinova, 2,6-dihydroxyisonicotinate, 4-pyridinecarboxylic acid, 2,6-dihydroxy, citrazinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo
PubChem CID 7425
IUPAC Name 6-hydroxy-2-oxo-1,2-dihydropyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=O)NC(O)=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 155.11
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Citrazinic Acid
EINECSNumber (5)-0744
RTECSNumber NS1400000
TSCA Yes