missing translation for 'onlineSavingsMsg'
Learn More

Citrate Buffer, pH 6.00, Ricca Chemical

Catalog No. 21050632
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21050632 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21050632 Supplier Ricca Chemical Company Supplier No. 2105.0632
Only null left

Specifications

Chemical Name or Material Citrate Buffer
Name Note pH 6.00
CAS 7732-18-5, 1310-73-2, 77-92-9
CAS Min % 96, 1.21%, 1.1%
Color Colorless
CAS Max % 100, 1.26%, 1.14%
Concentration 1X
For Use With (Application) Water Analysis
pH 6
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less