missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citrate Buffer, 0.2M buffer soln., pH 4.0, low endotoxin

Catalog No. AAJ67435AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67435AK 250 mL
AAJ67435AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67435AK Supplier Thermo Scientific Chemicals Supplier No. J67435AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6132-04-3 | C6H9Na3O9 | 294.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Citrate Buffer
Name Note low endotoxin
CAS 4-3-6132
Color Colorless
Concentration 0.2 M
pH 4.0
Physical Form Liquid
Quantity 250 mL
Molecular Formula C6H9Na3O9
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Molecular Weight (g/mol) 294.10
Show More Show Less

RUO – Research Use Only