missing translation for 'onlineSavingsMsg'
Learn More

Citral, cis + trans, 95%

CAS: 5392-40-5 | C10H16O | 152.24 g/mol

Supplier:  Thermo Scientific Chemicals A1616918

 View more versions of this product

Catalog No. AAA1616918


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Citral
5392-40-5
0.888
98°C (208°F)
1.488
50 g
14,2322
WTEVQBCEXWBHNA-JXMROGBWSA-N
(2E)-3,7-dimethylocta-2,6-dienal
638011
152.24
cis + trans
<-10°C
228°C to 229°C
C10H16O
MFCD00006997
1721871
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
CC(C)=CCC\C(C)=C\C=O
152.24
CHEBI:16980
95%

Chemical Identifiers

5392-40-5
152.24
WTEVQBCEXWBHNA-JXMROGBWSA-N
638011
(2E)-3,7-dimethylocta-2,6-dienal
C10H16O
MFCD00006997
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
CHEBI:16980
CC(C)=CCC\C(C)=C\C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H317
Causes skin irritation.
May cause an allergic skin reaction.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

EINECSNumber : 226-394-6

RTECSNumber : RG5075000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only