Learn More
Citral, cis + trans, 95%
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Supplier: Thermo Scientific Chemicals A1616922
| Quantity | 100 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C10H16O | |
| MFCD00006997 | |
| citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb | |
| CHEBI:16980 | |
| CC(C)=CCC\C(C)=C\C=O |
Specifications
| Citral | |
| 5392-40-5 | |
| 0.888 | |
| 98°C (208°F) | |
| 1.488 | |
| MFCD00006997 | |
| 14,2322 | |
| WTEVQBCEXWBHNA-JXMROGBWSA-N | |
| (2E)-3,7-dimethylocta-2,6-dienal | |
| 638011 | |
| 152.24 |
| cis + trans | |
| <-10°C | |
| 228°C to 229°C | |
| C10H16O | |
| 100 g | |
| 1721871 | |
| citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb | |
| CC(C)=CCC\C(C)=C\C=O | |
| 152.24 | |
| CHEBI:16980 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H317
Causes skin irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
H315-H317-H319
EINECSNumber : 226-394-6
RTECSNumber : RG5075000
TSCA : Yes
Recommended Storage : Keep cold