Learn More
Citraconic acid, 98+%
CAS: 498-23-7 | C5H6O4 | 130.10 g/mol
Supplier: Thermo Scientific Chemicals L0417818
Description
Citraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCitraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications
| Citraconic acid | |
| 88°C to 91°C | |
| C5H6O4 | |
| 50 g | |
| Hygroscopic | |
| 2-methylmaleate, citraconate 2-, z-2-methylbut-2-enedioate | |
| HNEGQIOMVPPMNR-IHWYPQMZSA-N | |
| (Z)-2-methylbut-2-enedioate | |
| 5461090 | |
| 130.1 |
| 498-23-7 | |
| 1.62 | |
| MFCD00078086 | |
| 1722679 | |
| 14,2321 | |
| Soluble in water. | |
| C\C(=C\C(O)=O)C(O)=O | |
| 130.10 | |
| CHEBI:30719 | |
| ≥98% |
Chemical Identifiers
| 498-23-7 | |
| 130.10 | |
| HNEGQIOMVPPMNR-IHWYPQMZSA-N | |
| 5461090 | |
| (Z)-2-methylbut-2-enedioate |
| C5H6O4 | |
| MFCD00078086 | |
| 2-methylmaleate, citraconate 2-, z-2-methylbut-2-enedioate | |
| CHEBI:30719 | |
| C\C(=C\C(O)=O)C(O)=O |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 207-858-7
RTECSNumber : GE6650000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only