missing translation for 'onlineSavingsMsg'
Learn More

Citraconic acid, 98+%

Catalog No. p-7044036
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0417809 10 g
AAL0417818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0417809 Supplier Thermo Scientific Chemicals Supplier No. L0417809
Only null left

CAS: 498-23-7 | C5H6O4 | 130.10 g/mol

Citraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citraconic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.

Chemical Identifiers

CAS 498-23-7
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.10
MDL Number MFCD00078086
InChI Key HNEGQIOMVPPMNR-IHWYPQMZSA-N
Synonym 2-methylmaleate, citraconate 2-, z-2-methylbut-2-enedioate
PubChem CID 5461090
ChEBI CHEBI:30719
IUPAC Name (Z)-2-methylbut-2-enedioate
SMILES C\C(=C\C(O)=O)C(O)=O

Specifications

Melting Point 88°C to 91°C
Density 1.62
Quantity 10 g
Beilstein 1722679
Sensitivity Hygroscopic
Merck Index 14,2321
Solubility Information Soluble in water.
Formula Weight 130.1
Percent Purity ≥98%
Chemical Name or Material Citraconic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-858-7
RTECSNumber GE6650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only